Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

4-Bromo-2-nitrophenol ≥98%

Glass bottle for solids, 5 g
Catalog # ALFAH26529.06
Irritant
4-Bromo-2-nitrophenol ≥98%
Catalog # ALFAH26529.06
CAS Number:  7693-52-9
4-Bromo-2-nitrophenol ≥98%
4-Bromo-2-nitrophenol ≥98%

Glass bottle for solids, 5 g

CAS Number:  7693-52-9
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 7693-52-9 Molecular Formula: C6H4BrNO3 Formula Weight: 218.00 Melting Point: 88-91° MDL No.: MFCD00082540

  • CAS Number:7693-52-9
  • Formula:C₆H₄BrNO₃
  • MDL Number:MFCD00082540
  • Molecular Weight:218.01 g/mol
  • Boiling Pt:259 °C (1013 hPa)
  • Melting Pt:88…91 °C
  • Storage Temperature:Ambient
  • EINECS:231-707-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications