Order Entry
Singapore
ContactUsLinkComponent
Ph-spear combination electrode with 1 m fixed cable and din plug, glass shaft, 3 x ceramic diaphragm, electrolyte kcl 3 mol/l, iodin e/iodide-reference system, a-glass, length 120 mm, 12/8 mm , -5...+80 c, 0...14 ph 1 * 1 items
Catalog # SCOI285113940
Supplier:  SI Analytics
Ph-spear combination electrode with 1 m fixed cable and din plug, glass shaft, 3 x ceramic diaphragm, electrolyte kcl 3 mol/l, iodin e/iodide-reference system, a-glass, length 120 mm, 12/8 mm , -5...+80 c, 0...14 ph 1 * 1 items
Catalog # SCOI285113940
Supplier:  SI Analytics