Titanium carbide, nanopowder
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Titanium carbide, nanopowder
Catalog # ALFA44922.14
Supplier: Thermo Fisher Scientific
CAS Number: 12070-08-5
Jump to:
Product Details & Documents
CAS No.: 12070-08-5 Molecular Formula: CTi Formula Weight: 59.91 Physical Form: 30-50nm APS Powder, S.A. 35-45m2/g Density: 4.93 Appearance: Black Melting Point: ca 3140° Boiling Point: 4820° UN No.: 3178 MDL No.: MFCD00011268
- CAS Number:12070-08-5
- Formula:CTi
- MDL Number:MFCD00011268
- Molecular Weight:59.89 g/mol
- Boiling Pt:4820 °C
- Melting Pt:∼ 3140 °C
- Density:4,93
- Storage Temperature:Ambient
- EINECS:235-120-4
- UN:3178
- ADR:4.1,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



