Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
COD standards
COD standards
Print
COD standards
COD standard 1000 mg/l, 1
Catalog # ORIOCODS01
Supplier:
Thermo Orion
COD standards
Catalog # ORIOCODS01
Supplier:
Thermo Orion
CAS Number:
Expand image
Return to Top
COD standards
COD standard 1000 mg/l, 1
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Inquire For Price
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Specifications
Pk:
1
Description:
COD standard 1000 mg/l
Specifications