Order Entry
Singapore
ContactUsLinkComponent
Phosphoglucose isomerase. (e.c.5.3.1.9) from baker's yeast crystalline suspension in 3.2 m ammonium sulfate activity: min. 300 units /mg protein unit definition: one unit converts one micromole of fructose-6-phosphate to glucose-6-phosphate per minut 1 * 1 KU
Catalog # ICNA0210098301
Supplier:  MP Biomedicals
Phosphoglucose isomerase. (e.c.5.3.1.9) from baker's yeast crystalline suspension in 3.2 m ammonium sulfate activity: min. 300 units /mg protein unit definition: one unit converts one micromole of fructose-6-phosphate to glucose-6-phosphate per minut 1 * 1 KU
Catalog # ICNA0210098301
Supplier:  MP Biomedicals