Microplate replicators
Replicator, 48-pin
BOEI140501
The Microplate replicator is a convenient, time-saving device for transferring small volumes of inoculum from microplates to other microplates or solid supports. Applications include replication of YACs and cosmid libraries, inoculation of filters for colony hybridisation, antibiotic sensitivity testing, and phage typing.
- Pins made of stainless steel for durability, autoclaving and flame sterilisation
- Economical alternative to robotic systems
- 48- and 96-pin models include an alcohol reservoir and cleaning pads for debris removal
- Template model available for 384-pin replicator
Note: It is recommended that 2 templates be used when replicating between two 384-well plates.
- Description:Replicator, 48-pin
- Seq:01