Order Entry
Singapore
ContactUsLinkComponent
tris-acetate/sds running buffer is used to resolve high molecular weight proteins (36-400 kda) under denaturing conditions on tris/a cetate gels 1 * 500 mL
Catalog # GENO786-198
Supplier:  G-Biosciences
tris-acetate/sds running buffer is used to resolve high molecular weight proteins (36-400 kda) under denaturing conditions on tris/a cetate gels 1 * 500 mL
Catalog # GENO786-198
Supplier:  G-Biosciences