Order Entry
Singapore
Orders LinkContactUsLinkComponent
Ethanol absolute ≥99.5%, TechniSolv®, pure

Ethanol absolute ≥99.5%, TechniSolv®, pure

Plastic container, Duty paid, 20 L
Catalog # VWRC83813.440_T
Flammable
Irritant
Ethanol absolute ≥99.5%, TechniSolv®, pure
Catalog # VWRC83813.440_T
Supplier:  
CAS Number:  64-17-5
Ethanol absolute ≥99.5%, TechniSolv®, pure
Ethanol absolute ≥99.5%, TechniSolv®, pure

Plastic container, Duty paid, 20 L

CAS Number:  64-17-5
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Ethanol absolute TechniSolv pure 1 * 10 L Catalog #VWRC83813.410

Product VWRC83813.410 is a replacement for VWRC83813.440_T but varies by Brand, Packaging or Quantity.

Ethanol absolute TechniSolv pure 1 * 5 L Catalog #VWRC83813.360

Product VWRC83813.360 is a replacement for VWRC83813.440_T but varies by Brand, Packaging or Quantity.

Product Details & Documents

Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.

Synonyms:  Ethyl alcohol
  • CAS Number:64-17-5
  • Formula:C₂H₅OH
  • MDL Number:MFCD00003568
  • Molecular Weight:46.07 g/mol
  • Merck Index:12,03806
  • Boiling Pt:78,3 °C (1013 hPa)
  • Melting Pt:–117 °C
  • Density:0,7895 g/cm³ (20 °C)
  • Flash Pt:12 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-578-6
  • UN:1170
  • ADR:3,II
  • REACH:01-2119457610-43
Specifications

Specification Test Results

  • Assay (V/V):
    Min. 99.5 %
  • Density (20/20):
    0.788 - 0.794
  • Water:
    Max. 0.5 %

Specifications

  • Pack type:
    Plastic container
  • Pk:
    20 L
  • Duty status:
    Duty paid

Specifications