Ethanol absolute ≥99.5%, TechniSolv®, pure
Plastic jerrican, Duty paid, 5 L
Ethanol absolute ≥99.5%, TechniSolv®, pure
Catalog # VWRC83813.360DP
CAS Number: 64-17-5
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Merck Index:12,03806
- Boiling Pt:78,3 °C (1013 hPa)
- Melting Pt:–117 °C
- Density:0,7895 g/cm³ (20 °C)
- Flash Pt:12 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-578-6
- UN:1170
- ADR:3,II
- REACH:01-2119457610-43
Documents
Certificates
Search by Lot Number10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
P20_VWRC83813.360_SPEC_EN.pdf P20_VWRC83813.410_SPEC_EN.pdf P20_VWRC83813.440_SPEC_EN.pdf spec_VWRC83813.360 spec_VWRC83813.410
Product specification
More
VWR Chemicals Big Packs and Bulks VWR Denatured Ethanol
Product Literature
Specifications
Specification Test Results
- Assay (V/V):Min. 99.5 %
- Density (20/20):0.788 - 0.794
- Water:Max. 0.5 %
Specifications
- Pack type:Plastic jerrican
- Pk:5 L
- Duty status:Duty paid



