Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
COD standards
Print
COD standards
COD standard 10000 mg/l, 475 mL
Catalog # ORIOCODS10
Supplier:
Thermo Orion
COD standards
Catalog # ORIOCODS10
Supplier:
Thermo Orion
CAS Number:
Return to Top
COD standards
COD standard 10000 mg/l, 475 mL
Specifications
Jump to:
Specifications
CAS Number:
Price
Inquire For Price
Log in
for availability
1 * 475 mL
-
+
Inquire For Price
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Specifications
Pk:
475 mL
Description:
COD standard 10000 mg/l
Specifications