Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Pipette, 25:0.1ml, graduated, class AS, DIN 12697 1 * 1 items
Catalog # HIRS110017037
Supplier:
HIRSCHMANN
Expand image
Pipette, 25:0.1ml, graduated, class AS, DIN 12697 1 * 1 items
Catalog # HIRS110017037
Supplier:
HIRSCHMANN