Order Entry
Singapore
Orders LinkContactUsLinkComponent
Ethyl isothiocyanate ≥97%

Ethyl isothiocyanate ≥97%

Glass bottle for solids, 500 g
Catalog # ALFAA19492.36
Toxic
Health
Corrosive
Flammable
Ethyl isothiocyanate ≥97%
Catalog # ALFAA19492.36
CAS Number:  542-85-8
Ethyl isothiocyanate ≥97%
Ethyl isothiocyanate ≥97%

Glass bottle for solids, 500 g

CAS Number:  542-85-8
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 97% CAS No.: 542-85-8 Molecular Formula: CH3CH2NCS Formula Weight: 87.14 Flash Point: 32°(89°F) Refractive Index: 1.5110 Odour: Pungent Melting Point: -6° Boiling Point: 130-132° Beilstein Reference: 1737705 Merck Reference: 14,3815 UN No.: 2929 MDL No.: MFCD00004820

  • CAS Number:542-85-8
  • Formula:C₃H₅NS
  • MDL Number:MFCD00004820
  • Molecular Weight:87.15 g/mol
  • Merck Index:14,3815
  • Boiling Pt:130…132 °C
  • Melting Pt:–6 °C
  • Density:0,995
  • Flash Pt:32 °C (89 °F)
  • Storage Temperature:Ambient
  • EINECS:208-831-2
  • UN:2929
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications