Order Entry
Singapore
Orders LinkContactUsLinkComponent
1,2-Dibromo-1-phenylethane ≥97%
Glass bottle for solids, 25 g
  ALFAB24990.14
1,2-Dibromo-1-phenylethane ≥97%
  ALFAB24990.14
 :  93-52-7
1,2-Dibromo-1-phenylethane ≥97%
1,2-Dibromo-1-phenylethane ≥97%
Glass bottle for solids, 25 g
 :  93-52-7
1 * 25 g
Purity: 97% CAS No.: 93-52-7 Molecular Formula: C8H8Br2 Formula Weight: 263.96 Melting Point: 70-74° Boiling Point: 139-140°/15mm UN No.: 3261 MDL No.: MFCD00000138
 :  (1,2-Dibromoethyl)benzene
  •  :93-52-7
  •  :C₆H₅CHBrCH₂Br
  •  :MFCD00000138
  •  :263.96 g/mol
  •  :139…140 °C (15 mmHg)
  •  :70…74 °C
  •  :1,74 g/cm³ (20 °C)
  •  :Ambient
  •  :202-253-4
  •  :3261
  •  :8,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g