Order Entry
Singapore
Orders LinkContactUsLinkComponent
1,1,3,3-Tetramethoxypropane ≥98%

1,1,3,3-Tetramethoxypropane ≥98%

Glass bottle for solids, 5.000 g
Catalog # ALFAA11418.0I
Flammable
1,1,3,3-Tetramethoxypropane ≥98%
Catalog # ALFAA11418.0I
CAS Number:  102-52-3
1,1,3,3-Tetramethoxypropane ≥98%
1,1,3,3-Tetramethoxypropane ≥98%

Glass bottle for solids, 5.000 g

CAS Number:  102-52-3
Price
Inquire For Price
Log in for availability
1 * 5.000 g
Product Details & Documents

Purity: 98+% CAS No.: 102-52-3 Molecular Formula: C7H16O4 Formula Weight: 164.20 Flash Point: 54°(129°F) Refractive Index: 1.4070 Boiling Point: 182-183° Beilstein Reference: 1700020 UN No.: 3271 MDL No.: MFCD00008488

Synonyms:  Malonaldehyde bis(dimethyl acetal)
  • CAS Number:102-52-3
  • Formula:C₇H₁₆O₄
  • MDL Number:MFCD00008488
  • Molecular Weight:164.2 g/mol
  • Boiling Pt:182…183 °C
  • Density:0,997
  • Flash Pt:54 °C (129 °F)
  • Storage Temperature:Ambient
  • EINECS:203-037-2
  • UN:3271
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g

Specifications