Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Flask volumetric, capacity 5l, with rim, DURAN, class A, tolerance 1.2ml, height 470mm, graduation color blue, conformity certified. 1 * 1 items
HIRS2800197_S
:
HIRSCHMANN
Flask volumetric, capacity 5l, with rim, DURAN, class A, tolerance 1.2ml, height 470mm, graduation color blue, conformity certified. 1 * 1 items
HIRS2800197_S
:
HIRSCHMANN