Order Entry
Singapore
ContactUsLinkComponent
Flask volumetric, capacity 200ml, class A, amber graduated, glass stoppered, tolerance 0.15ml, height 210mm, NS-ground 14/23, gradua tion color white, conformity certified. 1 * 1 items
Catalog # HIRS2640285_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 200ml, class A, amber graduated, glass stoppered, tolerance 0.15ml, height 210mm, NS-ground 14/23, gradua tion color white, conformity certified. 1 * 1 items
Catalog # HIRS2640285_S
Supplier:  HIRSCHMANN