Order Entry
Singapore
ContactUsLinkComponent
Flask volumetric, capacity 20ml, class A, amber graduated, glass stoppered, tolerance 0.04ml, height 110mm, NS-ground 10/19, graduat ion color white, conformity certified. 1 * 1 items
Catalog # HIRS2640265_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 20ml, class A, amber graduated, glass stoppered, tolerance 0.04ml, height 110mm, NS-ground 10/19, graduat ion color white, conformity certified. 1 * 1 items
Catalog # HIRS2640265_S
Supplier:  HIRSCHMANN