Pipette volumetric, capacity 3ml, graduation brown in mass, class AS, 2 marks, conformity certified. 1 * 1 items
Catalog # HIRS1340248_S
Pipette volumetric, capacity 3ml, graduation brown in mass, class AS, 2 marks, conformity certified. 1 * 1 items
Catalog # HIRS1340248_S