Pipette volumetric, capacity 15ml, class AS, blue graduation, tolerance 0.03ml, total length +-10mm:510mm, conformity certified. 1 * 1 items
Catalog # HIRS1341164_S
Pipette volumetric, capacity 15ml, class AS, blue graduation, tolerance 0.03ml, total length +-10mm:510mm, conformity certified. 1 * 1 items
Catalog # HIRS1341164_S