Order Entry
Singapore
ContactUsLinkComponent
Pipette volumetric, capacity 10ml, class AS, blue graduation, tolerance 0.02ml, total length +-10mm:440mm, conformity certified. 1 * 1 items
Catalog # HIRS1341160_S
Supplier:  HIRSCHMANN
undefined
Pipette volumetric, capacity 10ml, class AS, blue graduation, tolerance 0.02ml, total length +-10mm:440mm, conformity certified. 1 * 1 items
Catalog # HIRS1341160_S
Supplier:  HIRSCHMANN