Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

Spatulas, round, bent

210 mm, 12×5 mm
Catalog # BOCH32025
Supplier:   BOCHEM
Spatulas, round, bent
Catalog # BOCH32025
Supplier:  BOCHEM
Spatulas, round, bent
Spatulas, round, bent

210 mm, 12×5 mm

Price
$64.10
Log in for availability
1 * 5 items
Product Details & Documents

Stainless steel ¹⁸/₁₀, anti-magnetic. These spatulas with one rounded end can be used for a variety of applications.

  • Excellent heat resistance, autoclavable
  • Good resistance to chemicals
  • Easy and quick cleaning

Documents

Specifications
  • Length:
    210 mm
  • Seq:
    06
  • Spoon size:
    12×5 mm