Order Entry
Singapore
ContactUsLinkComponent
Comb, device,electrophoresis,gel,12cm length,with 25 teeth,1mm tooth thickness,8l sample volume with average gel thickness,teeth are precision machined from tough acrylic and the spine is made from thick PTFE to offer heat resistant stiffness,apollo 1 * 1 items
Catalog # BITX75.1214-MT225C
Supplier:  BIOTIX
Comb, device,electrophoresis,gel,12cm length,with 25 teeth,1mm tooth thickness,8l sample volume with average gel thickness,teeth are precision machined from tough acrylic and the spine is made from thick PTFE to offer heat resistant stiffness,apollo 1 * 1 items
Catalog # BITX75.1214-MT225C
Supplier:  BIOTIX