Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
New, competitively priced dispersing instrument for volumes of 0,03 to 100 ml. 1 * 1 items
Catalog # IKAA3240003
Supplier:
IKA
Expand image
New, competitively priced dispersing instrument for volumes of 0,03 to 100 ml. 1 * 1 items
Catalog # IKAA3240003
Supplier:
IKA