Order Entry
Singapore
ContactUsLinkComponent
The IKA LR-2.ST system is a modulary designed miniplant reactor system, planned and designed to simulate and optimize chemical reac tion processes as well as mixing, dispersion and homogenization processes at a model scale with a maximum volume of 2 1 * 1 items
Catalog # IKAA8016502
Supplier:  IKA
undefined
The IKA LR-2.ST system is a modulary designed miniplant reactor system, planned and designed to simulate and optimize chemical reac tion processes as well as mixing, dispersion and homogenization processes at a model scale with a maximum volume of 2 1 * 1 items
Catalog # IKAA8016502
Supplier:  IKA