Order Entry
Singapore
ContactUsLinkComponent
Quiet, economical laboratory stirrer with direct motor drive which can be infinitely adjusted by electronic means. For stirring subs tances of low to medium viscosity. 1 * 1 items
Catalog # IKAA3331402
Supplier:  IKA
Quiet, economical laboratory stirrer with direct motor drive which can be infinitely adjusted by electronic means. For stirring subs tances of low to medium viscosity. 1 * 1 items
Catalog # IKAA3331402
Supplier:  IKA