Order Entry
Singapore
Orders LinkContactUsLinkComponent
Methyl-2-methoxypropionate 98%

Methyl-2-methoxypropionate 98%

25 g
Catalog # ALFAL13527.14
Flammable
Irritant
Methyl-2-methoxypropionate 98%
Catalog # ALFAL13527.14
CAS Number:  17639-76-8
Methyl-2-methoxypropionate 98%
Methyl-2-methoxypropionate 98%

25 g

CAS Number:  17639-76-8
Price
$1,390.00
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 17639-76-8 Molecular Formula: C5H10O3 Formula Weight: 118.13 Flash Point: 36°(96°F) Refractive Index: 1.3970 Boiling Point: 43-44°/15mm Beilstein Reference: 1720819 UN No.: UN3272 MDL No.: MFCD00151664

Synonyms:  Methyl 2-methoxypropanoate
  • CAS Number:17639-76-8
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00151664
  • Molecular Weight:118.13 g/mol
  • Boiling Pt:43…44 °C (15 mmHg)
  • Flash Pt:36 °C (96 °F)
  • Storage Temperature:Ambient
  • EINECS:241-622-4
  • ADR:3,III
Specifications

Specifications

  • Pk:
    25 g

Specifications