Order Entry
Singapore
Orders LinkContactUsLinkComponent
General purpose spoons

General purpose spoons

With cylindrical rod (Ø 5 mm), 150 mm, 15×35 mm
Catalog # RSGA329.150
Supplier:   RSG
General purpose spoons
Catalog # RSGA329.150
Supplier:  RSG
General purpose spoons
General purpose spoons

With cylindrical rod (Ø 5 mm), 150 mm, 15×35 mm

Price
Inquire For Price
Log in for availability
1 * 5 items
Product Details & Documents

Double-ended spoons made from stainless steel.

  • Round handle (Ø 5 mm)
  • One spatula end, one spoon end
Specifications
  • Description:
    With cylindrical rod (Ø 5 mm)
  • Length:
    150 mm
  • Spoon size:
    15×35 mm