QUANTOFIX® Test Strips, Nitrate/Nitrite
Product Details & Documents
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready to use. They do not require additional accessories. The test strips are intended for single-use, maintenance or calibration are not required.
- Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips are for the rapid and reliable determination of nitrate and nitrite in solutions. The easy 'dip and read' test strips provide a reliable result within 1 minute.
Sets of three individually sealed test strips, pack of 50 sets. Without nitrite color scale, but with nitrite test pad.
Specifications
Specifications
- Chemical method:—
- Pk:1 KIT
- Description:QUANTOFIX® nitrate, test sets
- Dimensions:93×130×45 mm (packaging)
- Tests per kit:150 Tests
- Range:0 • 10 • 25 • 50 • 100 • 250 • 500 - mg/L - NO₃⁻
- Cat. No.:MANA913918
- Parameters:All reagents included
- Contents:50 sets of 3 individually sealed test strips each
- Platform:Semi-quantitative test strips
- Compliance:—
- Instrument:No
- Shelf life (months):6 (from production)
- Method:Nitrate - Modified Griess reaction (white → red-violet)
- Weight:241,88 g



