Order Entry
Singapore
Orders LinkContactUsLinkComponent
Absorbent papers

Absorbent papers

Absorbent paper roll, 600×500 mm, 0,22 mm
Catalog # MANA112030
Supplier:   MACHEREY-NAGEL
Absorbent papers
Catalog # MANA112030
Supplier:  MACHEREY-NAGEL
Absorbent papers
Absorbent papers

Absorbent paper roll, 600×500 mm, 0,22 mm

Price
Inquire For Price
Log in for availability
1 * 1 Roll
Product Details & Documents

Filter paper coated on one side with polyethylene for covering laboratory work benches absorbs spilt liquids.

Suitable for isotope and bacteriological laboratories as well as for chemical storerooms and cupboards.

Specifications
  • Description:
    Absorbent paper roll
  • L×W:
    600×500 mm
  • Seq:
    004
  • Thickness:
    0,22 mm