Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

Absorbent papers

Absorbent paper sheets, 480×600 mm, 0,22 mm
Catalog # MANA112000.1
Supplier:   MACHEREY-NAGEL
Absorbent papers
Catalog # MANA112000.1
Supplier:  MACHEREY-NAGEL
Absorbent papers
Absorbent papers

Absorbent paper sheets, 480×600 mm, 0,22 mm

Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents

Filter paper coated on one side with polyethylene for covering laboratory work benches absorbs spilt liquids.

Suitable for isotope and bacteriological laboratories as well as for chemical storerooms and cupboards.

Specifications
  • Description:
    Absorbent paper sheets
  • L×W:
    480×600 mm
  • Seq:
    002
  • Thickness:
    0,22 mm