Order Entry
Singapore
Orders LinkContactUsLinkComponent
Spatulas, round, bent
130 mm, 10×5 mm
  BOCH3200
 :   BOCHEM
Spatulas, round, bent
  BOCH3200
 :  BOCHEM
Spatulas, round, bent
Spatulas, round, bent
130 mm, 10×5 mm
$11.20
1 * 1 items
Stainless steel ¹⁸/₁₀, anti-magnetic. These spatulas with one rounded end can be used for a variety of applications.
  • Excellent heat resistance, autoclavable
  • Good resistance to chemicals
  • Easy and quick cleaning
  • Length:
    130 mm
  • Seq:
    01
  • Spoon size:
    10×5 mm