Methanol ≥99.5%, GPR RECTAPUR®
Plastic bottle, 2,5 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Merck Index:13,05984
- Boiling Pt:64,6 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0,7918 g/cm³ (20 °C)
- Flash Pt:11 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3 (6.1),II
- REACH:01-2119433307-44
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 225L Metal drum UN (ZPACE8010050) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic Jerrycan for Liquids DIN 42 UN (ZPACK13951065)
Specifications
Specification Test Results
- Assay:Min. 99.5 %
- Appearance:Clear colourless liquid
- IR Spectrum:Passes test
- Acidity:Max. 0.0002 meq/g
- Boiling point:64 - 65 °C
- Colouration:Max. 10 APHA
- Acetone:Max. 10 ppm
- Evaporation residue:Max. 50 ppm
- Water:Max. 0.2 %
Specifications
- Pack type:Plastic bottle
- Pk:2,5 L



