Order Entry
Singapore
Orders LinkContactUsLinkComponent
Lactic acid 90% ≥90% in aqueous solution, TECHNICAL

Lactic acid 90% ≥90% in aqueous solution, TECHNICAL

Glass bottle, 1 L
Catalog # VWRC20366.293
Corrosive
Supplier:   VWR Chemicals
Lactic acid 90% ≥90% in aqueous solution, TECHNICAL
Catalog # VWRC20366.293
Supplier:  VWR Chemicals
CAS Number:  50-21-5
Lactic acid 90% ≥90% in aqueous solution, TECHNICAL
Lactic acid 90% ≥90% in aqueous solution, TECHNICAL

Glass bottle, 1 L

CAS Number:  50-21-5
Price
$136.00
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid
  • CAS Number:50-21-5
  • Formula:H₃CCH(OH)COOH
  • MDL Number:MFCD00004520
  • Molecular Weight:90.08 g/mol
  • Boiling Pt:122 °C (20 hPa)
  • Melting Pt:53 °C
  • Density:1,11…1.21 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-018-0
Specifications

Specification Test Results

  • Assay:
    Min. 90 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications