Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Capillary column coated with BP10(1701) 30m x 0.32mm id, 0.5m film.(SGE 054265)
568-0384
:
Trajan Scientific and Medical
Capillary column coated with BP10(1701) 30m x 0.32mm id, 0.5m film.(SGE 054265)
568-0384
:
Trajan Scientific and Medical