Microplate 1536 well polystyrene Low Volume 2µl clear flat bottom non-treated without lid non-sterile 1 * 100 items
Catalog # CORN3850
Microplate 1536 well polystyrene Low Volume 2µl clear flat bottom non-treated without lid non-sterile 1 * 100 items
Catalog # CORN3850