Utility carts, MW series
MW703, 760×457×965 mm, 19,5 kg, 3
Catalog # METOMW703
Supplier: Metro International
Jump to:
Product Details & Documents
These steel or chrome plated carts consist of Super Erecta shelves with plastic split sleeves, two one-piece handles of matching finish, and designated castors with donut bumpers. These multipurpose trolleys are perfect for carrying out an enormous variety of tasks.
- Ergonomically designed for low effort transport
- Precise handling for manoeuvring in close quarters
- Sturdy aluminum corner castings firmly secure solid shelves to posts
- Stable, tough, simple and easy to clean
- MW carts use light-duty resilient castors
Specifications
- Description:MW703
- Seq:32
- W×D×H:760×457×965 mm
- Shelves:3
- Weight:19,5 kg



