Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Titanium(IV) methylphenoxide, tech. 1 * 10 g
Titanium(IV) methylphenoxide, tech. 1 * 10 g
Print
Titanium(IV) methylphenoxide, tech. 1 * 10 g
Catalog # ALFA89454.09
Certificates
SDS
Titanium(IV) methylphenoxide, tech. 1 * 10 g
Catalog # ALFA89454.09
Supplier:
CAS Number:
SDS
Certificates
Expand image
Return to Top
Titanium(IV) methylphenoxide, tech. 1 * 10 g
Add to cart
CAS Number:
Discontinued
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
SDS
msds_ALFA089454
Videos
Thermo Scientific Acroseal 25 Years