Wire baskets
Wire diameter 0,51 mm, 8 turns, wire leads 50 mm, 7x9 mm
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Made from Tungsten
- Wire Baskets for laboratory needs for holding high temperature samples.
CAS No.: 7440-33-7 MDL No.: MFCD00011461
For vacuum deposition
Documents
Specifications
- Ø×H:7x9 mm
- Description:Wire diameter 0,51 mm, 8 turns, wire leads 50 mm
- Seq:02



