Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

Insulating tubes, ceramic

0,8 mm, 1,6 mm, 200 mm
  ALFA31485.DR
Insulating tubes, ceramic
  ALFA31485.DR
Insulating tubes, ceramic

0,8 mm, 1,6 mm, 200 mm

 
 
   
1 * 200 mm
 

Pure Al₂O₃ Aluminium oxide AL23.

Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.

 
  • Tubing ext.Ø:
    1,6 mm
  • Tubing int.Ø:
    0,8 mm
  • Length:
    200 mm
  • Seq:
    29