Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

Insulating tubes, ceramic

0,5 mm, 1,0 mm, 100 mm
  ALFA31478.DM
Insulating tubes, ceramic
  ALFA31478.DM
Insulating tubes, ceramic

0,5 mm, 1,0 mm, 100 mm

 
 
   
1 * 100 mm
 

Pure Al₂O₃ Aluminium oxide AL23.

Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.

 
  • Tubing ext.Ø:
    1,0 mm
  • Tubing int.Ø:
    0,5 mm
  • Length:
    100 mm
  • Seq:
    12