Small flanges with hose nozzle
Small flanges, DN 25, Aluminium, 15 mm
VACU20662519
These flanges are made from aluminium or plastic.
- Fast, easy assembly and disassembly to/from pumps
- Safe, vacuum tight connection with optional centring and clamping rings
- A wide range of flanges with hose nozzles for various internal hose diameters are available
For connecting pumps to vacuum hoses.
- Tubing int.Ø:15 mm
- Description:Small flanges, DN 25
- Material:Aluminium
- Seq:08