Small flanges with hose nozzle
Small flanges, DN 25, Aluminium, 8 mm
Catalog # VACU20662516
Supplier: VACUUBRAND
Jump to:
Product Details & Documents
These flanges are made from aluminium or plastic.
- Fast, easy assembly and disassembly to/from pumps
- Safe, vacuum tight connection with optional centring and clamping rings
- A wide range of flanges with hose nozzles for various internal hose diameters are available
For connecting pumps to vacuum hoses.
Documents
MB_Kleinflansche_Schlauchwelle MB_Kleinflansche_Schlauchwelle_alt
Technical Drawing
Specifications
- Tubing int.Ø:8 mm
- Description:Small flanges, DN 25
- Material:Aluminium
- Seq:05