Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
0.53mm ID x 1µm film thickness x 25m length BP1 GC Capillary Column 1 * 1 items
SGEA054087
:
Trajan Scientific and Medical
0.53mm ID x 1µm film thickness x 25m length BP1 GC Capillary Column 1 * 1 items
SGEA054087
:
Trajan Scientific and Medical
Trajan Scientific GC Consumables