Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SGE SYRINGE 250µL WITH A GAS TIGHT PLUNGER AND 1/4-28 UNF SCREW THREAD FOR SHIMADZU TOC 2000 SERIES
Catalog # 548-1645
Supplier:
Trajan Scientific and Medical
Expand image
SGE SYRINGE 250µL WITH A GAS TIGHT PLUNGER AND 1/4-28 UNF SCREW THREAD FOR SHIMADZU TOC 2000 SERIES
Catalog # 548-1645
Supplier:
Trajan Scientific and Medical