Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SGE SYRINGE 25µL REMOVABLE NEEDLE IS 0.63 MM OD (23 GAUGE) 42 MM LONG AND HAS A CONE POINT STYLE. FOR MANUAL USE ON AGILENT AUTOSAMP LER.
Catalog # 549-0387
Supplier:
Trajan Scientific and Medical
Expand image
SGE SYRINGE 25µL REMOVABLE NEEDLE IS 0.63 MM OD (23 GAUGE) 42 MM LONG AND HAS A CONE POINT STYLE. FOR MANUAL USE ON AGILENT AUTOSAMP LER.
Catalog # 549-0387
Supplier:
Trajan Scientific and Medical