Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SGE SYRINGE 10µL REMOVABLE NEEDLE FOR SHIMADZU AOC9 AUTOSAMPLER NEEDLE IS STAINLESS STEEL 0.47 MM OD (26 GAUGE) 50 MM LONG WITH A CO NE POINT STYLE.
Catalog # 549-0407
Supplier:
Trajan Scientific and Medical
Expand image
SGE SYRINGE 10µL REMOVABLE NEEDLE FOR SHIMADZU AOC9 AUTOSAMPLER NEEDLE IS STAINLESS STEEL 0.47 MM OD (26 GAUGE) 50 MM LONG WITH A CO NE POINT STYLE.
Catalog # 549-0407
Supplier:
Trajan Scientific and Medical