Order Entry
Singapore
ContactUsLinkComponent
SGE SYRINGE 5µL FIXED NEEDLE NEEDLE IS STAINLESS STEEL 42 MM LONG 23 GAUGE (0.63MM OD) WITH CONE POINT STYLE. FOR MANUAL INJECTION O N AGILENT 7673/7683 EQUIPPED SYSTEMS
Catalog # 549-0427
SGE SYRINGE 5µL FIXED NEEDLE NEEDLE IS STAINLESS STEEL 42 MM LONG 23 GAUGE (0.63MM OD) WITH CONE POINT STYLE. FOR MANUAL INJECTION O N AGILENT 7673/7683 EQUIPPED SYSTEMS
Catalog # 549-0427