Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
SGE SYRINGE 5µL REMOVABLE NEEDLE NEEDLE IS STAINLESS STEEL 51 MM LONG 22 GAUGE (0.71 MM OD) WITH LC POINT STYLE
549-0454
:
Trajan Scientific and Medical
SGE SYRINGE 5µL REMOVABLE NEEDLE NEEDLE IS STAINLESS STEEL 51 MM LONG 22 GAUGE (0.71 MM OD) WITH LC POINT STYLE
549-0454
:
Trajan Scientific and Medical