Order Entry
Singapore
ContactUsLinkComponent
GLOVES CONFORM + : AMBIDEXTROUS SINGLE-USE LATEX GLOVES. PRODUCED UNDER ISO 9002. POWDER FREE. TEXTURED FINISH. NATURAL COLOUR. SIZ E 5-6.5
Catalog # 112-0976
Supplier:  Ansell
GLOVES CONFORM + : AMBIDEXTROUS SINGLE-USE LATEX GLOVES. PRODUCED UNDER ISO 9002. POWDER FREE. TEXTURED FINISH. NATURAL COLOUR. SIZ E 5-6.5
Catalog # 112-0976
Supplier:  Ansell