Storage microplates 384-well polypropylene, Corning®
Product Details & Documents
These 384-well polypropylene plates and blocks are built to the industry standard footprint.
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
They are automation-friendly and solvent-resistant. They feature round-, conical-, or v-bottoms for optimal liquid handling and a rounded square-well geometry for reduced wicking. Both plates and blocks are certified DNase- and RNase-free.
Ordering information: The 384-well plates are packaged 25 per pack, except for CORN3672 which is packaged 10 per pack. The 384-well blocks are packaged 5 per pack.
Documents
Certificates
Search by Lot NumberSpecifications
- Sterile:+
- Colour:Clear
- Description:384-well storage block, clear, round well bottom
- Seq:04
- W×D×H:127,76×85,48×27,81 mm
- Well volume:180 µl
Accessories
Product Information
- Catalog #SterileAvailabilityPrice
- Chemical resistant storage mat, PP, for 384-well deep well PP storage blockChemical resistant storage mat, PP, for 384-well deep well PP storage blockCORN3341-Log in for availability1 * 50 items - $1,680.00



